Would you like to log in again?
Clorox® Tilex® Soap Scum Remover, Essendant
Supplier: Essendant
Clorox® Tilex®
CLO35604EA
500032-888EA 14.44 USD InStock
500032-888
Clorox® Tilex® Soap Scum Remover, Essendant
Cleaning Agents Detergents and Surface Cleansers
Cleans and deodorizes glazed ceramic tile, plastic, chrome, fiberglass, glazed porcelain and other hard, nonporous surfaces.
  • Unscented liquid
  • Nonabrasive
  • Bactericidal and mildewstatic
Removes tough soap scum on contact; no scrubbing needed.
Certifications: EPA Registered.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty