Would you like to log in again?
Sealed Air Jiffy® TuffGard® Self-Seal Cushioned Mailer
Supplier: Sealed Air
Jiffy®
SEL49676 SEL37714 SEL49675 SEL37713 SEL37715 SEL49674 SEL10122 SEL37712 SEL49678 SEL24300
500009-358CT 68.8 USD InStock
500009-358 500016-964 500017-031 500017-032 500009-344 500009-362 500017-028 500017-026 500009-319 500017-027
Sealed Air Jiffy® TuffGard® Self-Seal Cushioned Mailer
Mailers and Shippers
Barrier Bubble® lining retains air better than alternative cushioning materials.
  • Co-extruded seams are heat sealed and burst proof
  • Self-seal closure features a tamper indicator
Polypropylene exterior is moisture, puncture and rip resistant.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty