Would you like to log in again?
Sheila Shine Stainless Steel Cleaner and Polish
Supplier: Essendant
SSI1EA SSI4EA SSI4CT SSI2EA
500021-768CT 201.91 USD InStock
500021-768 500032-958 500035-860 500032-954
Sheila Shine Stainless Steel Cleaner and Polish
Cleaning Agents Detergents and Surface Cleansers
Removes greasy film and water marks.
  • Acetone based
  • Unscented liquid
Resists fingerprints and streaking. Repels water; preserves surface against deterioration. Requires no rubbing or polishing.
In addition to stainless steel, this also works on Formica®, porcelain, fiberglass, enamel, leather and plastic.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty