Would you like to log in again?
Eco-Products® Renewable Resource Compostable Straws, Essendant
Supplier: Essendant
Eco-Products®
ECOEPST770
500025-970CT 382.31 USD InStock
500025-970
Eco-Products® Renewable Resource Compostable Straws, Essendant
Foodservice Supplies
Make the earth friendly choice with your break room supplies
Straws are made with corn-based NatureWorks® PLA. Odorless, non-allergenic and completely compostable under commercial conditions. You won't even know the difference, but you'll be making one.
Certifications: Compliant with ASTM D6400 Compostable Standard.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty