Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Innovera® Laser Cartridge, 7570A, Essendant LLC MS

Supplier: Essendant
Innovera®
IVR7570A
500029-022EA 140.48 USD InStock
500029-022
Innovera® Laser Cartridge, 7570A, Essendant LLC MS
Inks
Creates razor-sharp images and text on every page.

  • Compatible
  • Remanufactured
  • With 5% coverage

Resists fading so that the prints look just as good as the day made them. Easy to install, quick to replace, and simple to use.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty