Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Xerox® Solid Stick, 108R00723, 108R00724, 108R00725, 108R00726, 108R00727, 108R00727, 016213900, Essendant LLC MS

Supplier: Essendant
Xerox®
XER108R00726 XER108R00725 XER108R00723 XER108R00724
500015-048BX 256.38 USD InStock
500015-048 500015-049 500015-046 500015-047
Xerox® Solid Stick, 108R00723, 108R00724, 108R00725, 108R00726, 108R00727, 108R00727, 016213900, Essendant LLC MS
Inks
Drop-in loading of solid ink sticks is as easy as loading a stapler.

  • OEM and not remanufactured
  • With 5% coverage

You'll save space, because the packages of ink sticks are small enough to fit in a desk drawer. Solid ink sticks are safe and clean to handle.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty