Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Safco® Two-Shelf Wire Utility Cart

Supplier: Safco
Safco®
SAF5337BL
500004-587EA 272.78 USD InStock
500004-587
Safco® Two-Shelf Wire Utility Cart
Carts
Transports and stores materials safely and efficiently holding up to 200 lbs.

  • Rolls smoothly on four swivel casters
  • Mobile on four swivel 3" nylon casters

Shelves conveniently adjust in 1" increments. Clear plastic liner included for each shelf.

Certifications: GREENGUARD Indoor Air Quality Certified®.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty