Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
uni-ball® Signo 207™ Needle Point Roller Ball Pen
Supplier: Essendant
uni-ball® Signo 207™
UBC1736098 UBC1736097
500020-908DZ 25.8 USD InStock
500020-908 500020-909
uni-ball® Signo 207™ Needle Point Roller Ball Pen
Pens Ink Pens
The retractable pen features the Uni-Super Ink™ ink supply system that prevents from check and document fraud.
  • Needle point delivers precise writing every time
  • Soft textured grip provides a superior writing comfort and control
  • Medium point with 0.7 mm
  • With pocket clip and visible ink supply
Non-refillable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty