Would you like to log in again?
San Jamar® Tear-N-Dry Touchless Roll Towel Dispenser
Supplier: San Jamar
San Jamar®
SJMT1300TBK
500008-415EA 191.48 USD InStock
500008-415
San Jamar® Tear-N-Dry Touchless Roll Towel Dispenser
Wipe Dispensers
Hygienic touchless dispensing prevents germ spread
Just tear off towel and sensor activates to present another! Portion control provides 10” length every time. Dispenses any paper quality. Holds rolls up to 8” wide x 8” dia. and one 4” stub roll. Automatic transfer device allows complete usage of stub roll to eliminate waste. Lock prevents pilferage (waffle key incl.). Operates on four D alkaline batteries (sold separately); battery usage equivalent to dispensing 100, 800-ft. rolls. Break-resistant plastic.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty