Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
3M™ High-Capacity Maintenance Sorbent Mini-Boom
Supplier: 3M
MMMMMB312
500025-054CT 129.42 USD InStock
500025-054
3M™ High-Capacity Maintenance Sorbent Mini-Boom
Spill Control Materials
Designed for easily wrap around the base of equipment to help contain fluids that may leak or spill.
  • Prevents spread of fluids to adjacent work areas
  • Provides increased fluid-holding capacity so less sorbent is used
  • Permits simple reclamation for recycling of valuable liquids and reuse of sorbent
  • Minimizes sorbent waste for disposal
Caution: 3M products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations that accompany each item.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty