Would you like to log in again?
Crews® Safety Goggles
Supplier: MCR Safety
Crews®
CRW2230R
500012-786EA 15.29 USD InStock
500012-786
Crews® Safety Goggles
Goggles
Provides a wide, unobstructed field of vision and can be worn with most half-mask respirators.
Transparent PVC body permits a comfortable fit over glasses. Protects against impact, splash.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty