Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Rolodex™ Mesh Jumbo Clip Dish

Supplier: Rolodex
Rolodex™
ROL62562
500013-374EA 5.25 USD InStock
500013-374
Rolodex™ Mesh Jumbo Clip Dish
Desk Supply Holders
Wire Mesh clip holder; dia. 4 3/8" diameter, 2" height.

  • Saves space and improves efficiency
  • 50% more capacity than standard clip dish

Round holder stores paper clips, push pins and rubber bands; has 50% more capacity than standard clip dish.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty