Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Canon® Toner Cartridge, 1977B001, 1978B001, 1979B001, 1980B001, Essendant LLC MS

Supplier: Essendant
Canon®
CNM1977B001 CNM1979B001 CNM1978B001 CNM1980B001
500027-745EA 125.89 USD InStock
500027-745 500027-744 500027-747 500027-746
Canon® Toner Cartridge, 1977B001, 1978B001, 1979B001, 1980B001, Essendant LLC MS
Inks
Produces quality images over and over again.

  • 5% coverage
  • OEM and no remanufacturing available

Designed for optimal printing. Quick and easy installation means less downtime.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty