Would you like to log in again?
Memphis™ Ninja® X Gloves
Supplier: Essendant
Memphis™ Ninja®
CRWN9674XL CRWN9674L CRWN9674S CRWN9674M
500012-941PR 5.35 USD InStock
500012-941 500012-931 500012-925 500012-937
Memphis™ Ninja® X Gloves
Gloves
Premium durability and softness complement a bi-polymer coating on palm and fingertips for solid protection.
  • Gloves are made of nylon and silicone/spandex material
  • CE Score: 412X
These high-performance gloves are treated with Actifresh® to kill bacteria and promote longlasting freshness. Industry applications include manufacturing, fabrication, inspections, and packaging when maximizing dexterity and sense of touch is required.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty