Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lexmark™ Toner Cartridge, C736H1CG, C736H1KG, C736H1MG, C736H1YG, Essendant LLC MS

Supplier: Essendant
Lexmark™
LEXC736H1YG LEXC736H1MG LEXC736H1CG LEXC736H1KG
500027-664EA 603.92 USD InStock
500027-664 500027-663 500027-662 500027-661
Lexmark™ Toner Cartridge, C736H1CG, C736H1KG, C736H1MG, C736H1YG, Essendant LLC MS
Inks
Prints sharp blacks and smooth grays

  • 5% coverage
  • Return program

Uses toner optimally. Quick to install and simple to replace.

OEM, not remanufactured.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty