Would you like to log in again?
Tripp Lite Eight-Outlet Safety Surge Suppressor
Supplier: Tripp Lite
Tripp Lite
TLM825SA TRPTLM812SA
10144-278EA 133.67 USD InStock
10144-278 500026-086
Tripp Lite Eight-Outlet Safety Surge Suppressor
Power Supplies
Rugged surge suppressor provides reliable surge, spike and line noise protection for equipment in industrial, automotive and factory environments
Wide-spaced outlets accommodate multiple transformer plugs without blocking other outlets. Green LED confirms that connected equipment is being protected.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty