Would you like to log in again?
3M™ White Super Polish Floor Pads 4100
Supplier: 3M
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MMM08483 MMM08476
500039-354CT 53.72 USD InStock
500039-354 500039-351
3M™ White Super Polish Floor Pads 4100
Wipes Cleaning Kits and Pads
For buffing very soft finishes or for polishing soft waxes on wood floors.
Removes scuffs and black heel marks. Enhances floor appearance.
Used with rotary machine speed of 175 to 600 rpm.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty