Would you like to log in again?
San Jamar® Oceans® Tear-N-Dry Electronic Touchless Roll Towel Dispenser
Supplier: San Jamar
San Jamar® Oceans®
SJMT1390TBK
500008-416EA 191.48 USD InStock
500008-416
San Jamar® Oceans® Tear-N-Dry Electronic Touchless Roll Towel Dispenser
Wipe Dispensers
Sanitary touchless dispensing prevents germ spread.
  • Dispenses all qualities of paper without jamming
  • Break-resistant plastic
  • Lock prevents pilferage
Tear off one towel and the sensor activates to present another. Portion control provides 10” length every time. Holds rolls up to 8” wide x 8” dia. and one 4” stub roll. Automatic transfer device allows complete usage of stub roll to eliminate waste. Operates on four D alkaline batteries (sold separately); battery usage equivalent to dispensing 100, 800-ft. rolls.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty