Would you like to log in again?
Lathem® Time 7500E Totalizing Time Recorder, Essendant
Supplier: Essendant
Lathem®
LTH7500E
500043-487EA 712.18 USD InStock
500043-487
Lathem® Time 7500E Totalizing Time Recorder, Essendant
Record Keeping Forms
Automatic time recorder calculates time for up to 100 employees with atomic clock accuracy. Works for pay periods or in job mode.
  • Calculates actual or rounded time, supports automatic break deduction
  • Features automatic print alignment and sensing of front and back of cards
  • Internal lithium battery maintains time
  • Card totals and program settings for up to 3 years without power
  • Optional rechargable battery keeps unit fully usable up to 24 hours in event of power failure
Language: English, French, Spanish
For wall or desk mount
Minute increments: 1/100th
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty