Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tennsco Snap-Together Closed Starter Set, Essendant LLC MS

Supplier: Essendant
Tennsco
TNN128848PCSD TNN1276PCSD
500010-710EA 867.89 USD InStock
500010-710 500010-716
Tennsco Snap-Together Closed Starter Set, Essendant LLC MS
Furniture Shelves
Save 50% of the floor space used by conventional files.

  • For highly active files and records
  • All shelves are adjustable
  • Two 'L' uprights for the frame
  • Heavy-duty rolled steel construction with sand finish
  • Letter size file format

Provides a single freestanding unit or use with Add-On Units for a complete wall system. Units snap together without tools or fasteners. 200- to 300-lbs. load capacity per shelf with weight evenly distributed.

Ordering information: Order one Starter Set and any number of matching height Add-On Units.

Delivery information: Includes adjustable 12" deep shelves, shelf supports and five file dividers per shelf.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty