Would you like to log in again?
KIMBERLY-CLARK PROFESSIONAL® WYPALL® Waterless Hand Wipes
Supplier: Kimberly-Clark
KIMBERLY-CLARK PROFESSIONAL® WYPALL®
KCC91371CT KCC91371EA
500030-653EA 18.98 USD InStock
500030-653 500037-910
KIMBERLY-CLARK PROFESSIONAL® WYPALL® Waterless Hand Wipes
Wipes Disposable Wipes and Towels Presaturated Wipes
Premoistened wipes have orange peel extract cleansing agent.
  • Citrus scented
  • Alcohol-free formula
  • Removes grease, grime, ink and paint
Soothing ingredients aid in moisturizing skin. Dual textured, smooth and rough sides.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty