Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Clarity Mono Test, Clarity Diagnostics

Supplier: Clarity Diagnostics
Clarity
DTG-MONO
95064-004CS 4864.89 USD InStock
95064-004
Clarity Mono Test, Clarity Diagnostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
Clarity provides an easy-to-use diagnostic product that can quickly test for Mononucleosis. This CLIA Waived test is ideal for point-of-care testing that allows you to provide accurate results in no more than 8 minutes. Package contains reagent, controls, tubes, and cassettes for 15 tests.

  • Sensitivity > 99%
  • Specificity of 98.6%
  • Fast and easy test results in 8 minutes or less
  • No age restriction (can be used on children)
  • 18 month dating

Adding this test to your practice has the potential to create a new revenue stream for your practice while making your facility more efficient at caring for patient’s needs.

Certifications: CLIA waived for whole blood only, can be used with plasma/serum.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty