Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide, beads, Reagent Grade

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

0173-1KG 0173-5KG
97064-526CS 639.13 USD InStock
97064-526 97064-528
Sodium hydroxide, beads, Reagent Grade
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 323 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627
Order Now
Specification Test Results
Chloride = 0.05 %
Heavy Metals = 0.006 %
Potassium = 0.1 %
Purity 98.5 %
Sodium Carbonate = 0.90 %
Sulfate = 0.005 %

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty