Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium permanganate ACS

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Danger

0277-2.5KG
97065-026EA 247.12 USD InStock
97065-026
Potassium permanganate ACS
Potassium permanganate


Formula: KMnO₄
MW: 158.03 g/mol
Melting Pt: > 240 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1,II
Merck Index: 12,07824
Order Now
Specification Test Results
Chloride & Chlorate 0.005%
Insolubles = 0.2%
Purity = 99%
Sulfate = 0.02%

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty