Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenol red sodium salt ACS

Supplier: VWR
Synonyms: Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxidePhenol red water soluble3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt

0648-50G
97062-104EA 118.82 USD InStock
97062-104
Phenol red sodium salt ACS
Phenol red sodium salt
A pH indicator (Yellow to Red; range 6.8 - 8.4).


Formula: C₁₉H₁₃NaO₅S
MW: 376.35 g/mol
Melting Pt: 285 °C
Storage Temperature: Ambient
MDL Number: MFCD00066901
CAS Number: 34487-61-1
EINECS: 252-057-8
Merck Index: 12,07397
Order Now
Specification Test Results
Solubility (0.1%, Water) Pass
Visual Transition Interval (pH 6.8-8.2) Pass

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty