Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Riboflavine (vitamin B2), high purity

Supplier: VWR
Synonyms: Lactoflavin7,8-Dimethyl-10-((2S,3S,4R)-2,3,4,5-tetrahydroxypentyl)benzo[g]pteridine-2,4(3H,10H)-dioneVitamin B2Vitamine G(-)-Riboflavin

0744-250G
97062-302CS 570.58 USD InStock
97062-302
Riboflavine (vitamin B2), high purity
Riboflavine (Vitamin B2)


Formula: C₁₇H₂₀N₄O₆
MW: 376.37 g/mol
Melting Pt: 280 °C
Storage Temperature: Ambient
MDL Number: MFCD00005022
CAS Number: 83-88-5
EINECS: 201-507-1
Merck Index: 12,08367
Order Now
Specification Test Results
Ash = 0.3 %
Loss on Drying = 1.5 %
Lumiflavin (O.D.) = 0.025
Purity = 98 %
Specific Rotation (-135°) -( -115°)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty