Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

G418 disulfate, Ultrapure

Supplier: VWR
Danger
Synonyms: G-418 disulfateG-418 disulphateG418 disulfateG-418 sulphateG418 sulphateG-418 sulfateGeneticin®

E859-5G E859-1G
97063-058EA 96.63 USD InStock
97063-058 97063-060
G418 disulfate, Ultrapure
G418 disulphate
Selection agent for both prokaryotic and eukaryotic transfected cells. An aminoglycoside similar to gentamycin, G418 is toxic to bacterial, yeast, higher plant and mammalian cells, protozoans, and helminths.


Formula: C₂₀H₄₄N₄O₁₈S₂
MW: 692.72 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058314
CAS Number: 108321-42-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty