Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrogen peroxide 30% (w/w) ACS

Supplier: VWR
Danger

E882-500ML
97064-798CS 2742.95 USD InStock
97064-798
Hydrogen peroxide 30% (w/w) ACS
Hydrogen peroxide


Formula: H₂O₂
MW: 34.01 g/mol
Boiling Pt: ∼107 °C (1013 hPa)
Melting Pt: <0 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1,II
Order Now
Specification Test Results
Ammonium 0.0005%
Assay (H2O2) 29% - 32%
Chloride = 0.0003%
Color (APHA) = 10
Heavy Metals = 0.0001%
Iron = 0.00005%
Nitrate = 0.0002%
Phosphate = 0.0002%
Residue After Evaporation = 0.002%
Sulfate = 0.0005%
Titrable Acid = 0.0006 meq/g

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty