Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iodoacetamide ≥98%, Proteomics Grade

Supplier: VWR
Danger
Synonyms: 2-Iodoacetamide

M216-30G
97064-926CS 3868.06 USD InStock
97064-926
Iodoacetamide ≥98%, Proteomics Grade
Iodoacetamide


Formula: ICH₂CONH₂
MW: 184.96 g/mol
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 91.0 °C…93.0 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008028
CAS Number: 144-48-9
EINECS: 205-630-1
UN: 2928
ADR: 6.1,II
Order Now
Specification Test Results
Loss on Drying 1%
Melting Point 90°C - 95°C
Protease none detected
Purity 98 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty