Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Creatine phosphate disodium salt, high purity

Supplier: VWR
Synonyms: Sodium creatine phosphateSodium creatine phosphate dibasicN-Methyl-N-(N-phosphonocarbamimidoyl)glycine, disodium saltPhosphocreatine disodium salt

0271-25G
97061-328EA 293.93 USD InStock
97061-328
Creatine phosphate disodium salt, high purity
Creatine phosphate disodium salt


Formula: C₄H₈N₃O₅P·2Na
MW: 255.077 g/mol
MDL Number: MFCD00014351
CAS Number: 922-32-7
Order Now
Specification Test Results
ATP = 0.002 %
Inorganic Phosphate = 0.5 %
Purity (Enzymatic) 97 %
Water (KF) 20 % - 24 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty