Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

NaOH/SDS Lysis solution, Ultra Pure Grade

Supplier: VWR

J611-500ML
97064-814EA 172.13 USD InStock
97064-814
NaOH/SDS Lysis solution, Ultra Pure Grade
NaOH/SDS Lysis
An ultra pure reagent for alkaline lysis of bacterial cells used in standard DNA purification procedures.


Storage Temperature: Ambient
UN: 1824
ADR: 8,III
Order Now
Specification Test Results
Conductivity (10%, Water) @25C Passes Test
Titration 7.6 - 8.4 g/L

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty