Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(-)-Cystine USP

Supplier: VWR
Synonyms: (2R,2'R)-3,3'-Disulfanediylbis(2-aminopropanoic acid)(2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid)(H-Cys-OH)2(R)-(-)-Cystine

J993-100G
97063-470CS 1494.67 USD InStock
97063-470
L(-)-Cystine USP
L(-)-Cystine
White crystals or crystalline powder.


Formula: HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
MW: 240.3 g/mol
Boiling Pt: 428 °C (1013 hPa)
Melting Pt: 261…262 °C
Density: 1.677 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064228
CAS Number: 56-89-3
EINECS: 200-296-3
Merck Index: 13,02811
Order Now
Specification Test Results
Loss on Drying 0.2%
Residue on Ignition 0.1 %
Specific Rotation (-225°) - (-215°)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty