Would you like to log in again?
VWR® DNA Molecular Weight Markers, 1 kb Ladder
Supplier: Avantor
VWRVK181-1ML
75997-546EA 122.78 USD InStock
75997-546
VWR® DNA Molecular Weight Markers, 1 kb Ladder
Electrophoresis Reagents Molecular Weight Markers
11 fragments ranging from 500 to 10,000 bp for easy band identification
  • Easy band identification
  • Available pre-mixed with loading dye
11 fragments ranging from 500 to 10,000 bp for easy band identification. Also available pre-mixed with loading dye or with EZ-Vision® One DNA dye.
Also available pre-mixed with loading dye or with EZ-Vision® One DNA dye.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty