Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tween® 20 (Polysorbate) 10% in aqueous solution, Proteomics Grade peroxide-free

Supplier: VWR
Synonyms: Polyoxyethylene sorbitan monolaurate

M228-10ML-5PK
97063-980EA 166.39 USD InStock
97063-980
Tween® 20 (Polysorbate) 10% in aqueous solution, Proteomics Grade peroxide-free
Tween® 20 (Polysorbate)
  • Optimized for isolating proteins and sample preparation
  • Contains < 1ug of peroxide per ml
  • Convenient concentrations pre-measured in ampoules
  • Long-term stability

VWR’s high purity Pro-PureTM grade detergents are carefully packed under inert gas, making them extremely stable. The detergents are provided in ready-to-use 10% solutions that are packaged in 10 ml ampoules for optimal convenience.


Formula: C₅₈H₁₁₄O₂₆
MW: 1227.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty