Would you like to log in again?
MicrewLock® Microcentrifuge Tube with Tamper-Evident Screw Cap, Simport
Supplier: Simport
MicrewLock®
T335-6SPRTP T335-6SPRTP
89126-740CS 244.57 USD InStock
CA11033-400 89126-740
MicrewLock® Microcentrifuge Tube with Tamper-Evident Screw Cap, Simport
Tubes Microcentrifuge Tubes
Self-standing, polypropylene tube is ideal for applications requiring a tamper-evident seal, as well as for transport, freezer storage, and boiling.
  • Packaged in tamperproof resealable bags
  • Caps are supplied slightly screwed on
  • Printed graduations and white marking areas for sample identification
  • Sterilized by gamma radiation
Screw cap features an innovative tamper-evident locking ring, leakproof O-ring seal, and a flat top for writing and packaging applications.
Certifications: Certified RNase-, DNase-, DNA-, and pyrogen-free.
Caution: Caution: Do not use for storage in the liquid phase of liquid nitrogen.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty