Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium fluoride, Optical Grade

Supplier: Thermo Scientific Chemicals
Danger

11023-18 011023-18
AA11023-18EA 233 USD InStock
CAAA11023-18 AA11023-18
Sodium fluoride, Optical Grade
Sodium fluoride


Formula: NaF
MW: 41.99 g/mol
Boiling Pt: 1704 °C
Melting Pt: 993 °C
Density: 2.78
Storage Temperature: Ambient
MDL Number: MFCD00003524
CAS Number: 7681-49-4
EINECS: 231-667-8
UN: 1690
ADR: 6.1,III
Merck Index: 13,08691
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/30/16640630.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty