Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenylphosphine

Supplier: Thermo Scientific Chemicals
Danger

030155-06 30155-06
AA30155-06EA 410.99 USD InStock
CAAA30155-06 AA30155-06
Phenylphosphine
Phenylphosphine


Formula: C₆H₇P
Boiling Pt: 160 °C
Density: 1,00
Storage Temperature: Ambient
MDL Number: MFCD00002094
CAS Number: 638-21-1
EINECS: 211-325-4
UN: 2845
ADR: 4.2,I
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty