Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl glyoxime ≥99% ACS

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2,3-Butanedione dioxime Diacetyl dioxime Diacetyl glyoxime

033310-30 33310-18 33310-30
AA33310-30EA 170.99 USD InStock
AA33310-30 CAAA33310-30 CAAA33310-18
Dimethyl glyoxime ≥99% ACS
Dimethyl glyoxime


Formula: CH₃C(=NOH)C(=NOH)CH₃
MW: 116.12 g/mol
Boiling Pt: 53 °C (8 mmHg)
Melting Pt: ∼ 240 °C (dec.)
Density: 1.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002117
CAS Number: 95-45-4
EINECS: 202-420-1
Merck Index: 13,03272
Order Now
Specification Test Results
Insoluble in alcohol 0.05%
Residue after ignition 0.0005
Suitability for nickel determination Pass test
Melting point 240°

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty