Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrabutylgermane 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Tetra-n-butylgermaniumTetra-n-butylgermane

34119-14 034119-06 034119-03 34119-03 34119-06 034119-14
AA34119-03EA 38.69 USD InStock
CAAA34119-14 CAAA34119-03 AA34119-03 AA34119-14 CAAA34119-06 AA34119-06
Tetrabutylgermane 99%
Tetrabutylgermane


Formula: C₁₆H₃₆Ge
MW: 301.07 g/mol
Boiling Pt: 277…279 °C
Density: 0,934
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015224
CAS Number: 1067-42-1
EINECS: 213-929-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty