Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid standardised solution 5.0 N in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Hydrogen sulfateOil of vitriolVitriol brown oil

035613-K2 35613-K2
AA35613-K2EA 56.89 USD InStock
AA35613-K2 CAAA35613-K2
Sulfuric acid standardised solution 5.0 N in aqueous solution
Sulfuric acid


Formula: H₂SO₄
MW: 98.08 g/mol
Density: 1,146 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty