Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium bromide, crystal optic rectangle 38.5 mm x 19.5 mm x 4 mm, unpolished

Supplier: Thermo Scientific Chemicals
Warning

38409-KT 038409-KT
AA38409-KTEA 44.69 USD InStock
CAAA38409-KT AA38409-KT
Potassium bromide, crystal optic rectangle 38.5 mm x 19.5 mm x 4 mm, unpolished
Potassium bromide


Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435 °C (1013 hPa)
Melting Pt: 730 °C
Density: 2.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 12,07780
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty