Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride, crystal optic disc 13 mm x 1 mm, polished both sides

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: HaliteRock saltSalineSaltSea salt

38443-KT 038443-KT
AA38443-KTEA 101.26 USD InStock
AA38443-KT CAAA38443-KT
Sodium chloride, crystal optic disc 13 mm x 1 mm, polished both sides
Sodium chloride
Common Applications: Spectroscopy, IR window


Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty