Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetic acid ≥99.4%, Environmental Grade Plus

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Ethanoic acidGlacial acetic acidMethanecarboxylic acidVinegar acid

38739-AP 038739-AP
AA38739-APEA 666.99 USD InStock
CAAA38739-AP AA38739-AP
Acetic acid ≥99.4%, Environmental Grade Plus
Acetic acid


Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118.1 °C
Melting Pt: 16.6 °C
Density: 1.049
Flash Pt: 40 °C (104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
REACH: 01-2119475328-30
https://us-legacy.vwr.com/stibo/web/std.lang.all/31/15/723115.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty