Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium orthovanadate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: tri-Potassium vanadium tetraoxide

39502-14 039502-14 039502-22 39502-22
AA39502-22EA 870.99 USD InStock
CAAA39502-22 CAAA39502-14 AA39502-22 AA39502-14
Potassium orthovanadate ≥99.9% (metals basis)
Potassium orthovanadate


Formula: K₃O₄V
MW: 232.23 g/mol
Melting Pt: 1300 °C
Storage Temperature: Ambient
MDL Number: MFCD00210642
CAS Number: 14293-78-8
EINECS: 238-220-6
UN: 3285
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty