Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphorus(III) fluoride 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Phosphorus trichloride

39525-18 039525-18
AA39525-18EA 1910.99 USD InStock
AA39525-18 CAAA39525-18
Phosphorus(III) fluoride 99%
Phosphorus(III) fluoride


Formula: Cl₃P
MW: 87.97 g/mol
Boiling Pt: –101.5 °C
Melting Pt: –151.5 °C
Density: 3.907
Storage Temperature: Ambient
MDL Number: MFCD00040325
CAS Number: 7719-12-2
EINECS: 232-008-7
UN: 1955
ADR: 2.3,I
Merck Index: 12,07515
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty