Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Carbon disulfide ≥99.9% ACS

Supplier: Thermo Scientific Chemicals
Danger

039785-AP 039785-K3 39785-AY
AA39785-APEA 120.95 USD InStock
AA39785-AP AA39785-K3 AA39785-AY
Carbon disulfide ≥99.9% ACS
Carbon disulphide
Electronic vacuum tubes, in the manufacturing of carbon tetrachloride, rayon, soil disinfectants, as a solvent


Formula: CS₂
MW: 76.14 g/mol
Boiling Pt: 46 °C
Melting Pt: –112 °C
Density: 1.263
Flash Pt: –30 °C (–22 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011321
CAS Number: 75-15-0
EINECS: 200-843-6
UN: 1131
ADR: 3,I
Merck Index: 13,01821
https://us-legacy.vwr.com/stibo/web/std.lang.all/53/94/785394.jpg
Order Now
Specification Test Results
Color (APHA) 10
Evaporation residue 0.002%
Hydrogen sulfide (H2S) Pass test (limit about 1.5ppm)
Sulfur dioxide (SO2) Pass test (limit about 2.5ppm)
H2O 0.05%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty