Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cobalt selenide ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Cobalt(II) selenide

040224-18 40224-18
AA40224-18EA 220.99 USD InStock
CAAA40224-18 AA40224-18
Cobalt selenide ≥99% (metals basis)
Cobalt selenide


Formula: CoSe
MW: 137.89 g/mol
Melting Pt: 1055 °C
Density: 7,650
Storage Temperature: Ambient
MDL Number: MFCD00016020
CAS Number: 1307-99-9
EINECS: 215-155-1
UN: 3283
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty