Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Deuterium bromide 48% (w/w) in D₂O (99% D) (Isotopic)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Hydrogen bromide-D1

42408-18 42408-09 042408-18 042408-09
AA42408-18EA 283.99 USD InStock
CAAA42408-18 CAAA42408-09 AA42408-18 AA42408-09
Deuterium bromide 48% (w/w) in D₂O (99% D) (Isotopic)
Deuterium bromide
NMR Solvent


Formula: DBr
MW: 81.9 g/mol
Boiling Pt: 126 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00044232
CAS Number: 13536-59-9
EINECS: 236-894-6
UN: 1788
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty