Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Strontium titanate substrate 5x5x0.5 mm, polished both sides 110 orientation

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Synonyms: Strontium titanate Strontium titanate (IV) Strontium titanium oxide

42669-KU 42669-KT 042669-KT
AA42669-KTEA 152.99 USD InStock
AA42669-KT CAAA42669-KU CAAA42669-KT
Strontium titanate substrate 5x5x0.5 mm, polished both sides 110 orientation
Strontium titanate


Formula: O₃SrTi
Melting Pt: 2080 °C
Density: 5.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049554
CAS Number: 12060-59-2
EINECS: 235-044-1
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty