Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Strontium titanate substrate 5x5x1 mm, polished both sides 110 orientation

Supplier: Thermo Scientific Chemicals
Synonyms: Strontium titanate Strontium titanate (IV) Strontium titanium oxide

42671-KT 42671-KU 042671-KT
AA42671-KTEA 90.49 USD InStock
AA42671-KT CAAA42671-KU CAAA42671-KT
Strontium titanate substrate 5x5x1 mm, polished both sides 110 orientation
Strontium titanate


Formula: O₃SrTi
Melting Pt: 2080 °C
Density: 5.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049554
CAS Number: 12060-59-2
EINECS: 235-044-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty